Stampa…

Val-Cit-PAB-OH ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: (S)-2-[(S)-2-Amino-3-methylbutanamido]-N-[4-(hydroxymethyl)phenyl]-5-ureidopentanamide

V0155-100MG V0155-25MG
TCIAV0155-100MGEA 240 EUR
TCIAV0155-100MG TCIAV0155-25MG
Val-Cit-PAB-OH ≥98.0% (by HPLC)
Val-Cit-PAB-OH
Formula: C₁₈H₂₉N₅O₄
Peso molecolare: 379,46 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD22381209
CAS Number: 159857-79-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 17.50 to 19.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR