Stampa…

Veratramine ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (2S,3R,5S)-2-((S)-1-((3S,6aR,11aS,11bR)-3-Hydroxy-10,11b-dimethyl-2,3,4,6,6a,11,11a,11b-octahydro-1H-benzo[a]fluoren-9-yl)ethyl)-5-methylpiperidin-3-ol

V0148-10MG V0148-50MG
TCIAV0148-10MGEA 31 EUR
TCIAV0148-10MG TCIAV0148-50MG
Veratramine ≥98.0% (by HPLC)
Veratramine
Formula: C₂₇H₃₉NO₂
Peso molecolare: 409,61 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00468124
CAS Number: 60-70-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,09952

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 208.0 to 212.0 deg-C
Specific rotation [a]20/D -64.0 to -68.0 deg (C=0.5, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR