trans-4-(4-Vinylcyclohexyl)benzonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-(trans-4-Vinylcyclohexyl)benzonitrile, 1-Cyano-4-(trans-4-vinylcyclohexyl)benzene
V0135-1G
V0135-5G
TCIAV0135-1GEA
48
EUR
TCIAV0135-1G
TCIAV0135-5G
trans-4-(4-Vinylcyclohexyl)benzonitrile ≥98.0% (by GC)
trans-4-(4-Vinylcyclohexyl)benzonitrile
Formula:
C₁₅H₁₇N Peso molecolare: 211,31 g/mol |
MDL Number:
MFCD11053490 CAS Number: 96184-42-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...