Vinylsulphonic acid ≥97.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₂H₄O₃S Flash Pt: 159°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD09743544 CAS Number: 1184-84-5 UN: 3265 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Purity (Bromination Method) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...