Stampa…

Verapamil hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Pericolo

V0118-25G V0118-5G
TCIAV0118-25GEA 154 EUR
TCIAV0118-25G TCIAV0118-5G
Verapamil hydrochloride ≥98.0% (by HPLC, titration analysis)
Verapamil hydrochloride
Formula: C₂₇H₃₈N₂O₄
MDL Number: MFCD00055208
CAS Number: 152-11-4
EINECS: 205-800-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,09950
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4d7ca8d7b36086b4cd16e79c8fe04dd051a3096&vid=f4d7ca8d7b36086b4cd16e79c8fe04dd051a3096&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR