Stampa…

Vinpocetine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (41S,13aS)-Ethyl 13a-ethyl-2,3,41,5,6,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate

V0115-100MG V0115-1G
TCIAV0115-100MGEA 26 EUR
TCIAV0115-100MG TCIAV0115-1G
Vinpocetine ≥98.0% (by HPLC, titration analysis)
Vinpocetine
Formula: C₂₂H₂₆N₂O₂
Peso molecolare: 350,46 g/mol
MDL Number: MFCD00211233
CAS Number: 42971-09-5
EINECS: 256-028-0
Merck Index: 14,09991
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50276f07890af5b438d9d90f67caae53bd0e0719&vid=50276f07890af5b438d9d90f67caae53bd0e0719&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C
Specific rotation [a]20/D +115.0 to +120.0 deg (C=2, Pyridine)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR