Stampa…

Venlafaxine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride

V0110-1G V0110-5G
TCIAV0110-1GEA 59 EUR
TCIAV0110-1G TCIAV0110-5G
Venlafaxine hydrochloride ≥98.0% (by HPLC, titration analysis)
Venlafaxine hydrochloride
Formula: C₁₇H₂₇NO₂
Peso molecolare: 313,87 g/mol
Punto di fusione: 217°C
MDL Number: MFCD03658865
CAS Number: 99300-78-4
Merck Index: 14,09946

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR