Stampa…

Vidarabine monohydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: (2R,3S,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol hydrate, Ara-A, Adenine 9-beta-D-Arabinofuranoside, 9-(beta-D-Arabinofuranosyl)adenine

V0098-1G V0098-5G
TCIAV0098-1GEA 88 EUR
TCIAV0098-1G TCIAV0098-5G
Vidarabine monohydrate ≥98.0% (by HPLC, titration analysis)
Vidarabine monohydrate
Formula: C₁₀H₁₃N₅O₄·H₂O
Peso molecolare: 285.26 g/mol
Punto di fusione: 253°C
MDL Number: MFCD00150980
CAS Number: 24356-66-9
Merck Index: 14,09976
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=071ac1102f77cf71b732e4903911b52c345a3078&vid=edad27186e4877bdcc01cb4e0b730394eb474589&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Specific rotation [a]20/D -2.0 to -4.0 deg (C=1, DMF)
Drying loss 5.0 to 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR