sulpho-EMCS ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Sulpho-N-Succinimidyl 6-Maleimidocaproate sodium salt, 6-Maleimidocaproic acid sulpho-N-Succinimidyl ester sodium salt
U0122-25MG
U0122-100MG
TCIAU0122-25MGEA
138
EUR
TCIAU0122-25MG
TCIAU0122-100MG
sulpho-EMCS ≥95.0% (by HPLC)
sulpho-EMCS
Formula:
C₁₄H₁₅N₂NaO₉S Peso molecolare: 410,33 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD01310272 CAS Number: 215312-86-0 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 6.50 to 7.10 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...