Stampa…

(±)-Sulphinpyrazone ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,2-Diphenyl-4-(2-(phenylsulphinyl)ethyl)pyrazolidine-3,5-dione, sulphinpyrazone, 1,2-Diphenyl-4-[2-(phenylsulphinyl)ethyl]pyrazolidine-3,5-dione

U0114-5G U0114-25G
TCIAU0114-5GEA 52 EUR
TCIAU0114-5G TCIAU0114-25G
(±)-Sulphinpyrazone ≥98.0% (by HPLC, titration analysis)
(±)-Sulphinpyrazone
Formula: C₂₃H₂₀N₂O₃S
Peso molecolare: 404,484 g/mol
MDL Number: MFCD00057279
CAS Number: 57-96-5
EINECS: 200-357-4
Merck Index: 14,08949

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR