Ulipristal Acetate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
CDB-2914, (11beta)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione
U0102-250MG
U0102-50MG
TCIAU0102-250MGEA
637
EUR
TCIAU0102-250MG
TCIAU0102-50MG
Ulipristal Acetate ≥98.0% (by HPLC, titration analysis)
Ulipristal Acetate
Formula:
C₃₀H₃₇NO₄ Peso molecolare: 475,63 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00899035 CAS Number: 126784-99-4 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 184.0 to 188.0 deg-C |
Specific rotation [a]20/D | +173 to +183 deg (C=1, CHCL3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...