Stampa…

Undecafluorohexanoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

U0067-25G U0067-5G
TCIAU0067-25GEA 255 EUR
TCIAU0067-25G TCIAU0067-5G
Undecafluorohexanoic acid ≥98.0% (by GC, titration analysis)
Undecafluorohexanoic acid
Formula: C₆HF₁₁O₂
Punto di ebollizione: 157°C
MDL Number: MFCD00198040
CAS Number: 307-24-4
EINECS: 206-196-6
UN: 3265
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR