TU-3 ≥98.0% (by HPLC) for organic electronics
Fornitore: TCI
|
Attenzione
|
T3924-100MG
T3924-250MG
TCIAT3924-100MGEA
430
EUR
TCIAT3924-100MG
TCIAT3924-250MG
TU-3 ≥98.0% (by HPLC) for organic electronics
TU-3
Formula:
C₅₂H₆₀N₆S₄ |
CAS Number:
1681007-44-2 |
Risultati del test di specificazione
Appearance | Green to Dark green powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Electron Mobility (muFET) | min. 0.50 cm2/Vs (cPVP Si/SiO2 substrate) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...