Stampa…

3-(Trichlorosilyl)propyl 2-Bromo-2-methylpropanoate ≥95.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Bromo-2-methylpropionic acid 3-(Trichlorosilyl)propyl ester

T3844-1G T3844-5G
TCIAT3844-1GEA 188 EUR
TCIAT3844-1G TCIAT3844-5G
3-(Trichlorosilyl)propyl 2-Bromo-2-methylpropanoate ≥95.0% (by GC)
3-(Trichlorosilyl)propyl 2-Bromo-2-methylpropanoate
Formula: C₇H₁₂BrCl₃O₂Si
Peso molecolare: 342,51 g/mol
CAS Number: 688359-84-4
UN: 2987
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR