Stampa…

4-(Trifluoromethyl)benzylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Aminomethylbenzotrifluoride hydrochloride, 4-(Trifluoromethyl)benzylammonium chloride, [4-(Trifluoromethyl)phenyl]methanamine hydrochloride

T3836-1G T3836-5G
TCIAT3836-1GEA 33 EUR
TCIAT3836-1G TCIAT3836-5G
4-(Trifluoromethyl)benzylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
4-(Trifluoromethyl)benzylamine hydrochloride
Formula: C₈H₈F₃N·HCl
Peso molecolare: 211,61 g/mol
Punto di fusione: 167°C
MDL Number: MFCD07780751
CAS Number: 3047-99-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR