4-(Trifluoromethyl)anilinium chloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Aminobenzotrifluoride HCl, 4-Aminobenzotrifluoride hydrochloride, α,α,α-Trifluoro-p-toluidine HCl, α,α,α-Trifluoro-p-toluidine hydrochloride, 4-(Trifluoromethyl)aniline hydrochloride
T3833-1G
T3833-5G
TCIAT3833-1GEA
27
EUR
TCIAT3833-1G
TCIAT3833-5G
4-(Trifluoromethyl)anilinium chloride ≥98.0% (by HPLC, titration analysis)
4-(Trifluoromethyl)anilinium chloride
Formula:
C₇H₇ClF₃N Peso molecolare: 197,59 g/mol Punto di fusione: 202°C |
MDL Number:
MFCD00007897 CAS Number: 90774-69-9 EINECS: 292-679-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...