Stampa…

4,4,5,5-Tetramethyl-2-(4-phenoxyphenyl)-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxybenzene, Phenoxyphenyl-4-boronic acid Pinacol ester

T3831-1G T3831-5G
TCIAT3831-1GEA 20 EUR
TCIAT3831-1G TCIAT3831-5G
4,4,5,5-Tetramethyl-2-(4-phenoxyphenyl)-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
4,4,5,5-Tetramethyl-2-(4-phenoxyphenyl)-1,3,2-dioxaborolane
Formula: C₁₈H₂₁BO₃
Peso molecolare: 296,17 g/mol
MDL Number: MFCD06795691
CAS Number: 269410-26-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR