Stampa…

3',4',5'-Trifluoroacetophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(3,4,5-Trifluorophenyl)ethan-1-one

T3825-5G T3825-25G
TCIAT3825-5GEA 47 EUR
TCIAT3825-5G TCIAT3825-25G
3',4',5'-Trifluoroacetophenone ≥98.0% (by GC)
3',4',5'-Trifluoroacetophenone
Formula: C₈H₅F₃O
Peso molecolare: 174,12 g/mol
MDL Number: MFCD00083533
CAS Number: 220141-73-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR