Stampa…

4-[1,2,4]Triazol-4-ylbenzoic acid ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(4H-1,2,4-Triazol-4-yl)benzoic acid, 4-(p-Carboxyphenyl)-1,2,4-triazole

T3788-1G T3788-200MG
TCIAT3788-1GEA 212 EUR
TCIAT3788-1G TCIAT3788-200MG
4-[1,2,4]Triazol-4-ylbenzoic acid ≥98.0% (by HPLC)
4-[1,2,4]Triazol-4-ylbenzoic acid
Formula: C₉H₇N₃O₂
Peso molecolare: 189,18 g/mol
MDL Number: MFCD06654839
CAS Number: 157069-48-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR