2,4,4-Trimethylpentan-2-amine hydroiodide ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,1,3,3-Tetramethylbutylamine Hydroiodide, tert-Octylamine Hydroiodide, tert-Octylammonium iodide
T3785-1G
T3785-5G
TCIAT3785-1GEA
24
EUR
TCIAT3785-1G
TCIAT3785-5G
2,4,4-Trimethylpentan-2-amine hydroiodide ≥97.0% (by titrimetric analysis)
2,4,4-Trimethylpentan-2-amine hydroiodide
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...