Stampa…

(Triphenylmethyl)thionyl Imide ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Tritylthionyl Imide, TrNSO

T3773-1G T3773-5G
TCIAT3773-1GEA 75 EUR
TCIAT3773-1G TCIAT3773-5G
(Triphenylmethyl)thionyl Imide ≥97.0% (by GC)
(Triphenylmethyl)thionyl Imide
Formula: C₁₉H₁₅NOS
Peso molecolare: 305,40 g/mol
MDL Number: MFCD32062442
CAS Number: 503596-47-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 96.0 to 100.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR