Stampa…

2-(p-Tolyl)pyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

T3759-200MG T3759-1G
TCIAT3759-200MGEA 61 EUR
TCIAT3759-200MG TCIAT3759-1G
2-(p-Tolyl)pyrimidine ≥98.0% (by GC)
2-(p-Tolyl)pyrimidine
Formula: C₁₁H₁₀N₂
Peso molecolare: 170,22 g/mol
MDL Number: MFCD08701658
CAS Number: 77232-13-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR