Stampa…

2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,4,6-Trimethyl-2,4,6-trivinyl-1,3,5,2,4,6-trioxatrisilinane

T3745-25G T3745-5G
TCIAT3745-25GEA 99 EUR
TCIAT3745-25G TCIAT3745-5G
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane ≥95.0% (by GC)
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane
Formula: C₉H₁₈O₃Si₃
Peso molecolare: 258,50 g/mol
Punto di ebollizione: 80°C
MDL Number: MFCD00023111
CAS Number: 3901-77-7
EINECS: 223-458-5

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 0.9690 to 0.9790
Refractive index n20/D 1.4240 to 1.4280

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR