Stampa…

Triclopyr ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: [(3,5,6-Trichloro-2-pyridinyl)oxy]acetic acid

T3742-25G T3742-5G
TCIAT3742-25GEA 197 EUR
TCIAT3742-25G TCIAT3742-5G
Triclopyr ≥96.0% (by GC, titration analysis)
Triclopyr
Formula: C₇H₄Cl₃NO₃
Peso molecolare: 256,46 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00072514
CAS Number: 55335-06-3
EINECS: 259-597-3
UN: 3077
ADR: 9,III
Merck Index: 14,09656

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR