Stampa…

Tazobactam ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (2S,3S,5R)-3-((1H-1,2,3-Triazol-1-yl)methyl)-3-methyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, Tazobactam acid

T3732-1G T3732-5G
TCIAT3732-1GEA 48 EUR
TCIAT3732-1G TCIAT3732-5G
Tazobactam ≥97.0% (by HPLC, titration analysis)
Tazobactam
Formula: C₁₀H₁₂N₄O₅S
Peso molecolare: 300,289 g/mol
MDL Number: MFCD00867002
CAS Number: 89786-04-9
Merck Index: 14,09083

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +162 to +167 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR