Stampa…

Tectorigenin ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 4',5,7-Trihydroxy-6-methoxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-6-methoxychromone

T3729-50MG T3729-10MG
TCIAT3729-50MGEA 356 EUR
TCIAT3729-50MG TCIAT3729-10MG
Tectorigenin ≥98.0% (by HPLC)
Tectorigenin
Formula: C₁₆H₁₂O₆
Peso molecolare: 300,27 g/mol
MDL Number: MFCD00597094
CAS Number: 548-77-6
Merck Index: 14,09107

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 62.10 to 64.60 %
Melting point 228.0 to 232.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR