Stampa…

4-Nitrophenylboronic acid pinacol ester ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 4-Nitrophenylboronic acid Pinacol ester, 1-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Nitrobenzeneboronic acid pinacol ester

T3710-1G T3710-5G
TCIAT3710-1GEA 26 EUR
TCIAT3710-1G TCIAT3710-5G
4-Nitrophenylboronic acid pinacol ester ≥98.0% (by GC)
4-Nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BNO₄
Peso molecolare: 249,07 g/mol
MDL Number: MFCD02179437
CAS Number: 171364-83-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR