3-(Trifluoromethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
4,4,5,5-Tetramethyl-2-[3-(trifluoromethyl)phenyl]-1,3,2-dioxaborolane, 3-(Trifluoromethyl)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)benzene
Formula:
C₁₃H₁₆BF₃O₂ Peso molecolare: 272,0712 g/mol |
MDL Number:
MFCD05863925 CAS Number: 325142-82-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...