2,4,6-Tri-tert-butylpyrimidine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,4,6-Tri-tertbutylpyrimidine
T3693-1G
T3693-5G
TCIAT3693-1GEA
31
EUR
TCIAT3693-1G
TCIAT3693-5G
2,4,6-Tri-tert-butylpyrimidine ≥98.0% (by GC)
2,4,6-Tri-tert-butylpyrimidine
Formula:
C₁₆H₂₈N₂ Peso molecolare: 248,41 g/mol |
MDL Number:
MFCD03094045 CAS Number: 67490-21-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...