Tubastatin A hydrochloride ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
N-Hydroxy-4-[[2-methyl-3,4-dihydro-1H-pyrido[4,3-b]indol-5(2H)-yl]methyl]benzamide hydrochloride, N-Hydroxy-4-((2-methyl-1,2,3,4-tetrahydro-5H-pyrido[4,3-b]indol-5-yl)methyl)benzamide hydrochloride, Tubastatin A HCl
T3687-100MG
T3687-25MG
TCIAT3687-100MGEA
389
EUR
TCIAT3687-100MG
TCIAT3687-25MG
Tubastatin A hydrochloride ≥97.0% (by HPLC)
Tubastatin A hydrochloride
Formula:
C₂₀H₂₁N₃O₂·HCl Peso molecolare: 371,87 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD20488052 CAS Number: 1310693-92-5 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 10.70 to 11.90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...