Temozolomide acid ≥98.0% (by HPLC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₆H₅N₅O₃ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD12032160 CAS Number: 113942-30-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...