Stampa…

Tris(ethyl Acetoacetato)aluminum(III) ≥97.0% (by titrimetric analysis) (contains ca. 2% 2-Butanol at maximum)

Fornitore: TCI

T3681-25G T3681-500G
TCIAT3681-25GEA 19 EUR
TCIAT3681-25G TCIAT3681-500G
Tris(ethyl Acetoacetato)aluminum(III) ≥97.0% (by titrimetric analysis) (contains ca. 2% 2-Butanol at maximum)
Tris(ethyl Acetoacetato)aluminum(III)
Formula: C₁₈H₂₇AlO₉
MDL Number: MFCD01317537
CAS Number: 15306-17-9
EINECS: 239-343-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR