Stampa…

2,3,5,6-Tetrachloro-4-(methylsulphonyl)pyridine ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

T3670-1G T3670-5G
TCIAT3670-1GEA 122 EUR
TCIAT3670-1G TCIAT3670-5G
2,3,5,6-Tetrachloro-4-(methylsulphonyl)pyridine ≥95.0% (by GC)
2,3,5,6-Tetrachloro-4-(methylsulphonyl)pyridine
Formula: C₆H₃Cl₄NO₂S
MDL Number: MFCD00054553
CAS Number: 13108-52-6
EINECS: 236-035-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 146.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR