(+)-Taxifolin ≥95.0% (by HPLC, titration analysis)
Fornitore: TCI
T3666-200MG
T3666-1G
TCIAT3666-200MGEA
74
EUR
TCIAT3666-200MG
TCIAT3666-1G
(+)-Taxifolin ≥95.0% (by HPLC, titration analysis)
(+)-Taxifolin
Formula:
C₁₅H₁₂O₇ Punto di fusione: 242°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00006845 CAS Number: 480-18-2 EINECS: 207-543-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % (calcd.on dried substance) |
Specific rotation [a]20/D | +19.0 to +23.0 deg (C=1, methanol) |
Water | max. 10.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...