Stampa…

Tetrabutylammonium toluene-4-sulphonate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetrabutylammonium p-Toluenesulphonate, Tetrabutylammonium 4-methylbenzenesulphonate

T3660-25G T3660-5G
TCIAT3660-25GEA 141 EUR
TCIAT3660-25G TCIAT3660-5G
Tetrabutylammonium toluene-4-sulphonate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium toluene-4-sulphonate
Formula: C₂₃H₄₃NO₃S
Peso molecolare: 413,6574 g/mol
MDL Number: MFCD00043227
CAS Number: 7182-86-7
EINECS: 230-548-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR