Stampa…

Trifluoroacetaldehyde hydrate (contains Total ca. 35% Water)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Trifluoroacetaldehyde hydrate, 2,2,2-Trifluoro-1,1-ethanediol

T3654-5G T3654-25G
TCIAT3654-5GEA 33 EUR
TCIAT3654-5G TCIAT3654-25G
Trifluoroacetaldehyde hydrate (contains Total ca. 35% Water)
Trifluoroacetaldehyde hydrate
Formula: C₂H₃F₃O₂
Peso molecolare: 116,039 g/mol
MDL Number: MFCD00167359
CAS Number: 421-53-4
UN: 1989
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b32cf9e09165ae463aa034a44d1a94928ce7347&vid=3b32cf9e09165ae463aa034a44d1a94928ce7347&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Water 32.0 to 37.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR