Stampa…

Tributyl(ethyl)phosphonium Diethyl Phosphate ≥96.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tributyl(ethyl)phosphonium diethylphosphate

T3641-5G T3641-25G
TCIAT3641-5GEA 59 EUR
TCIAT3641-5G TCIAT3641-25G
Tributyl(ethyl)phosphonium Diethyl Phosphate ≥96.0% (by titrimetric analysis)
Tributyl(ethyl)phosphonium Diethyl Phosphate
Formula: C₁₈H₄₂O₄P₂
Peso molecolare: 384,48 g/mol
MDL Number: MFCD17018975
CAS Number: 20445-94-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f50317a40c79d73a4756dae0c16ed28a1a44cd43&vid=f50317a40c79d73a4756dae0c16ed28a1a44cd43&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR