Stampa…

Tirofiban hydrochloride monohydrate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-(Butylsulfonyl)-O-[4-(4-piperidinyl)butyl]-L-tyrosine hydrochloride monohydrate, MK-383 HCl, (S)-2-(Butylsulphonamido)-3-(4-(4-(piperidin-4-yl)butoxy)phenyl)propanoic acid hydrochloride hydrate

T3640-250MG T3640-50MG
TCIAT3640-250MGEA 439 EUR
TCIAT3640-250MG TCIAT3640-50MG
Tirofiban hydrochloride monohydrate ≥98.0% (by HPLC)
Tirofiban hydrochloride monohydrate
Formula: C₂₂H₃₉ClN₂O₆S
Peso molecolare: 495,07 g/mol
Punto di fusione: 132°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD07368623
CAS Number: 150915-40-5
Merck Index: 14,09464

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.00 to 6.00 %
Specific rotation [a]20/D -12.0 to -16.0 deg (C=1, MeOH)
Water 3.1 to 4.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR