Stampa…

2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI

T3638-200MG T3638-1G
TCIAT3638-200MGEA 40 EUR
TCIAT3638-200MG TCIAT3638-1G
2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine ≥98.0% (by GC, titration analysis)
2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine
Formula: C₈H₅F₃N₄
MDL Number: MFCD26403657
CAS Number: 219508-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f4313bab922ed837fcc336f268ce1a964af8cee&vid=1f4313bab922ed837fcc336f268ce1a964af8cee&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 157.0 to 161.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR