Trihexyl(tetradecyl)phosphonium chloride ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
T3623-5G
T3623-25G
TCIAT3623-5GEA
33
EUR
TCIAT3623-5G
TCIAT3623-25G
Trihexyl(tetradecyl)phosphonium chloride ≥97.0% (by titrimetric analysis)
Trihexyl(tetradecyl)phosphonium chloride
Formula:
C₃₂H₆₈ClP Peso molecolare: 519,31 g/mol |
MDL Number:
MFCD04112746 CAS Number: 258864-54-9 UN: 1760 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (Nonaqueous Titration) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...