Stampa…

Triflusal ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Acetoxy-4-(trifluoromethyl)benzoic acid, Acetyl-4-(trifluoromethyl)salicylic acid, Drisgen

T3601-250MG T3601-1G
TCIAT3601-250MGEA 244 EUR
TCIAT3601-250MG TCIAT3601-1G
Triflusal ≥98.0% (by titrimetric analysis)
Triflusal
Formula: C₁₀H₇F₃O₄
Peso molecolare: 248,16 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00866793
CAS Number: 322-79-2
EINECS: 206-297-5
Merck Index: 14,09688
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=423d38f7e15b9fcb015d1976628d86e957dce799&vid=423d38f7e15b9fcb015d1976628d86e957dce799&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR