Stampa…

Tyrphostin RG 14620 ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 3-(3,5-Dichlorophenyl)-2-(pyridin-3-yl)acrylonitrile

T3576-25MG T3576-100MG
TCIAT3576-25MGEA 160 EUR
TCIAT3576-25MG TCIAT3576-100MG
Tyrphostin RG 14620 ≥98.0% (by HPLC)
Tyrphostin RG 14620
Formula: C₁₄H₈Cl₂N₂
Peso molecolare: 275,132 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00236445
CAS Number: 136831-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d69bf9eed259d52591a1be0f5f0ae88031c1d56b&vid=d69bf9eed259d52591a1be0f5f0ae88031c1d56b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.5 to 10.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR