Stampa…

RG-13022 ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3-(3,4-Dimethoxyphenyl)-2-(pyridin-3-yl)acrylonitrile, RG13022, Tyrphostin RG 13022

T3575-25MG T3575-100MG
TCIAT3575-25MGEA 167 EUR
TCIAT3575-25MG TCIAT3575-100MG
RG-13022 ≥98.0% (by GC)
RG-13022
Formula: C₁₆H₁₄N₂O₂
Peso molecolare: 266,3 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00236444
CAS Number: 136831-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae41b62f37e952da7213404e86ad1ae08f39ae8&vid=9a3f55a5632097f1393565c538bb4307a059a47e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR