Trimetilamina ∼13% ∼2 mol/L in acetonitrile
Fornitore: TCI
|
Pericolo
|
Formula:
C₃H₉N Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00008327 CAS Number: 75-50-3 EINECS: 200-875-0 UN: 3286 ADR: 3,II Merck Index: 14,09710 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Neutralization titration) | 13.0 to 15.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...