Tyrphostin AG 835 ≥98.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₈H₁₆N₂O₃ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00209855 CAS Number: 133550-37-5 |
|
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | +93.0 to +98.0 deg (C=0.5,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...