Trihexyl(tetradecyl)phosphonium dicyanamide ≥93.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Tetradecyltrihexylphosphonium dicyanamide
T3534-5G
T3534-25G
TCIAT3534-5GEA
33
EUR
TCIAT3534-5G
TCIAT3534-25G
Trihexyl(tetradecyl)phosphonium dicyanamide ≥93.0% (by total nitrogen basis)
Trihexyl(tetradecyl)phosphonium dicyanamide
Formula:
C₃₄H₆₈N₃P Peso molecolare: 549,90 g/mol |
MDL Number:
MFCD04117699 CAS Number: 701921-71-3 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Purity (with Total Nitrogen) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...