3-(Trifluoromethoxy)benzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
3-(Trifluoromethoxy)benzene-1-sulphonyl chloride
T3508-1G
T3508-5G
TCIAT3508-1GEA
23
EUR
TCIAT3508-1G
TCIAT3508-5G
3-(Trifluoromethoxy)benzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
3-(Trifluoromethoxy)benzenesulphonyl chloride
Formula:
C₇H₄ClF₃O₃S Peso molecolare: 260,62 g/mol |
MDL Number:
MFCD01091016 CAS Number: 220227-84-9 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...