Tyrphostin A23 ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(3,4-Dihydroxybenzylidene)malononitrile
T3503-25MG
T3503-100MG
TCIAT3503-25MGEA
148
EUR
TCIAT3503-25MG
TCIAT3503-100MG
Tyrphostin A23 ≥98.0% (by GC)
Tyrphostin A23
Formula:
C₁₀H₆N₂O₂ Peso molecolare: 186,1 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00133899 CAS Number: 118409-57-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...