Stampa…

Tetrakis(trimethylsiloxy)silane ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetrakis(trimethylsilyl) Orthosilicate, Tetrakis(trimethylsilyloxy)silane, Tetrakis(trimethylsilyl) orthosilicate

T3494-25ML T3494-5ML
TCIAT3494-25MLEA 104 EUR
TCIAT3494-25ML TCIAT3494-5ML
Tetrakis(trimethylsiloxy)silane ≥98.0% (by GC)
Tetrakis(trimethylsiloxy)silane
Formula: C₁₂H₃₆O₄Si₅
Peso molecolare: 384,85 g/mol
Punto di ebollizione: 220°C
Flash Pt: 76°C
MDL Number: MFCD00051587
CAS Number: 3555-47-3
EINECS: 222-613-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR