Stampa…

Demethyltexasin ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Demethyltexasin, 6,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6,7-Trihydroxyisoflavone, 6,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 6,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone

T3473-1G T3473-200MG
TCIAT3473-1GEA 302 EUR
TCIAT3473-1G TCIAT3473-200MG
Demethyltexasin ≥95.0% (by HPLC, titration analysis)
Demethyltexasin
Formula: C₁₅H₁₀O₅
Peso molecolare: 270,24 g/mol
MDL Number: MFCD00016953
CAS Number: 17817-31-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR